Assay Details
| Gene Symbol: cpeb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351720.1 | NP_001338649.1 | 4 - 5 | 1516 | 65 | |
| NM_200981.2 | NP_957275.2 | 4 - 5 | 1468 | 65 | ||
| XM_005173113.4 | XP_005173170.2 | 5 - 6 | 1568 | 65 | ||
| XM_009291021.3 | XP_009289296.1 | 5 - 6 | 1898 | 65 | ||
| XM_017358999.2 | XP_017214488.1 | 5 - 6 | 1568 | 65 | ||
| XM_017359000.2 | XP_017214489.1 | 4 - 5 | 1544 | 65 | ||
| GenBank mRNA | BC055522.1 | - | 4 - 5 | 1468 | 65 | |
| BC165168.1 | - | 3 - 4 | 1211 | 65 | ||
| EF033337.1 | - | 3 - 4 | 1663 | 65 | ||
More Information
Set Membership: |
|

cpeb4a
Genomic Map