Assay Details
| Gene Symbol: cpeb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351720.1 | NP_001338649.1 | 7 - 8 | 1836 | 96 | |
| NM_200981.2 | NP_957275.2 | 7 - 8 | 1788 | 96 | ||
| XM_005173113.4 | XP_005173170.2 | 8 - 9 | 1888 | 96 | ||
| XM_009291021.3 | XP_009289296.1 | 8 - 9 | 2218 | 96 | ||
| GenBank mRNA | BC055522.1 | - | 7 - 8 | 1788 | 96 | |
| BC165168.1 | - | 6 - 7 | 1531 | 96 | ||
| EF033337.1 | - | 6 - 7 | 1983 | 96 | ||
| EH535381.1 | - | 2 - 3 | 646 | 96 | ||
More Information
Set Membership: |
|

cpeb4a
Genomic Map