Assay Details
| Gene Symbol: tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200985.2 | NP_957279.2 | 4 - 5 | 438 | 64 | |
| GenBank mRNA | BC055146.1 | - | 4 - 5 | 379 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tube1| Gene Symbol: tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200985.2 | NP_957279.2 | 4 - 5 | 438 | 64 | |
| GenBank mRNA | BC055146.1 | - | 4 - 5 | 379 | 64 | |
Set Membership: |
|