Assay Details
| Gene Symbol: prkacba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201023.1 | NP_957317.1 | 2 - 3 | 342 | 110 | |
| GenBank mRNA | BC053227.1 | - | 2 - 3 | 342 | 110 | |
| BC165279.1 | - | 2 - 3 | 276 | 110 | ||
More Information
Set Membership: |
|

prkacba
Genomic Map