Assay Details
| Gene Symbol: pgam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201024.1 | NP_957318.1 | 1 - 2 | 506 | 104 | |
| GenBank mRNA | BC053127.1 | - | 1 - 2 | 506 | 104 | |
| BC152224.1 | - | 1 - 2 | 517 | 104 | ||
| BC171458.1 | - | 1 - 2 | 478 | 104 | ||
| BC171460.1 | - | 1 - 2 | 478 | 104 | ||
More Information
Set Membership: |
|

pgam2
Genomic Map