Assay Details
| Gene Symbol: srp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201096.3 | NP_957390.1 | 2 - 3 | 240 | 94 | |
| GenBank mRNA | BC049522.1 | - | 2 - 3 | 235 | 94 | |
| EB763853.1 | - | 2 - 3 | 231 | 94 | ||
| EH541182.1 | - | 2 - 3 | 261 | 94 | ||
More Information
Set Membership: |
|

srp9
Genomic Map