Assay Details
| Gene Symbol: pik3r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201143.2 | NP_957437.2 | 2 - 3 | 517 | 79 | |
| XM_005166277.4 | XP_005166334.1 | 2 - 3 | 270 | 79 | ||
| XM_021476737.1 | XP_021332412.1 | 5 - 6 | 623 | 79 | ||
| GenBank mRNA | BC045386.1 | - | 2 - 3 | 529 | 79 | |
| BC081552.1 | - | 2 - 3 | 517 | 79 | ||
| BC153435.1 | - | 2 - 3 | 529 | 79 | ||
| BC155253.1 | - | 2 - 3 | 533 | 79 | ||
| BC164683.1 | - | 2 - 3 | 253 | 79 | ||
More Information
Set Membership: |
|

pik3r3b
Genomic Map