Assay Details
| Gene Symbol: pik3r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201143.2 | NP_957437.2 | 4 - 5 | 796 | 78 | |
| XM_005166277.4 | XP_005166334.1 | 4 - 5 | 549 | 78 | ||
| XM_021476737.1 | XP_021332412.1 | 7 - 8 | 902 | 78 | ||
| GenBank mRNA | BC045386.1 | - | 4 - 5 | 808 | 78 | |
| BC081552.1 | - | 4 - 5 | 796 | 78 | ||
| BC153435.1 | - | 4 - 5 | 808 | 78 | ||
| BC155253.1 | - | 4 - 5 | 812 | 78 | ||
| BC164683.1 | - | 4 - 5 | 532 | 78 | ||
| BC171614.1 | - | 4 - 5 | 691 | 78 | ||
More Information
Set Membership: |
|

pik3r3b
Genomic Map