Assay Details
| Gene Symbol: pik3r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201143.2 | NP_957437.2 | 6 - 7 | 1066 | 79 | |
| XM_005166277.4 | XP_005166334.1 | 6 - 7 | 819 | 79 | ||
| XM_021476737.1 | XP_021332412.1 | 9 - 10 | 1172 | 79 | ||
| GenBank mRNA | BC045386.1 | - | 6 - 7 | 1078 | 79 | |
| BC081552.1 | - | 6 - 7 | 1066 | 79 | ||
| BC153435.1 | - | 6 - 7 | 1078 | 79 | ||
| BC155253.1 | - | 6 - 7 | 1082 | 79 | ||
| BC164683.1 | - | 6 - 7 | 802 | 79 | ||
| BC171614.1 | - | 6 - 7 | 961 | 79 | ||
More Information
Set Membership: |
|

pik3r3b
Genomic Map