Assay Details
| Gene Symbol: pik3r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201143.2 | NP_957437.2 | 9 - 10 | 1494 | 77 | |
| GenBank mRNA | BC045386.1 | - | 9 - 10 | 1506 | 77 | |
| BC081552.1 | - | 9 - 10 | 1494 | 77 | ||
| BC153435.1 | - | 9 - 10 | 1506 | 77 | ||
| BC164683.1 | - | 9 - 10 | 1230 | 77 | ||
| BC171614.1 | - | 9 - 10 | 1389 | 77 | ||
More Information
Set Membership: |
|

pik3r3b
Genomic Map