Assay Details
| Gene Symbol: nme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201195.1 | NP_957489.1 | 4 - 5 | 545 | 88 | |
| XM_017353700.2 | XP_017209189.1 | 3 - 4 | 536 | 88 | ||
| GenBank mRNA | BC049030.1 | - | 4 - 5 | 545 | 88 | |
| BC153640.1 | - | 3 - 4 | 373 | 88 | ||
More Information
Set Membership: |
|

nme4
Genomic Map