Assay Details
| Gene Symbol: grinab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308611.1 | NP_001295540.1 | 3 - 4 | 544 | 61 | |
| XM_005158451.1 | XP_005158508.1 | 4 - 5 | 683 | 61 | ||
| GenBank mRNA | BC053253.1 | - | 3 - 4 | 449 | 61 | |
| BC152278.1 | - | 3 - 4 | 458 | 61 | ||
| EH541500.1 | - | 3 - 4 | 598 | 61 | ||
More Information
Set Membership: |
|

grinab
Genomic Map