Assay Details
| Gene Symbol: eif3ja | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201214.2 | NP_957508.2 | 7 - 8 | 715 | 135 | |
| GenBank mRNA | BC055247.1 | - | 7 - 8 | 699 | 135 | |
More Information
Set Membership: |
|

eif3ja
Genomic Map