Assay Details
| Gene Symbol: chrnb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201220.1 | NP_957514.1 | 4 - 5 | 619 | 101 | |
| GenBank mRNA | AY256909.1 | - | 4 - 5 | 619 | 101 | |
| BC162133.1 | - | 4 - 5 | 595 | 101 | ||
| BC162137.1 | - | 4 - 5 | 595 | 101 | ||
More Information
Set Membership: |
|

chrnb3a
Genomic Map