Assay Details
| Gene Symbol: cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201290.1 | NP_958447.1 | 3 - 4 | 384 | 66 | |
| XM_005161447.2 | XP_005161504.1 | 4 - 5 | 738 | 66 | ||
| GenBank mRNA | BC044393.1 | - | 3 - 4 | 384 | 66 | |
| BC071416.1 | - | 3 - 4 | 421 | 66 | ||
| BC155176.1 | - | 4 - 5 | 745 | 66 | ||
More Information
Set Membership: |
|

cct6a
Genomic Map