Assay Details
| Gene Symbol: rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201457.1 | NP_958865.1 | 5 - 6 | 698 | 57 | |
| GenBank mRNA | BC042327.1 | - | 5 - 6 | 698 | 57 | |
| BC071335.1 | - | 5 - 6 | 708 | 57 | ||
| BC152281.1 | - | 5 - 6 | 674 | 57 | ||
| BC164808.1 | - | 5 - 6 | 608 | 57 | ||
More Information
Set Membership: |
|

rfc3
Genomic Map