Assay Details
| Gene Symbol: hdlbpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201474.1 | NP_958882.1 | 22 - 23 | 3179 | 119 | |
| XM_005165938.4 | XP_005165995.1 | 22 - 23 | 3215 | 119 | ||
| XM_005165939.4 | XP_005165996.1 | 22 - 23 | 3240 | 119 | ||
| GenBank mRNA | BC051153.1 | - | 22 - 23 | 3179 | 119 | |
More Information
Set Membership: |
|

hdlbpa
Genomic Map