Assay Details
| Gene Symbol: hdlbpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201474.1 | NP_958882.1 | 3 - 4 | 245 | 79 | |
| XM_005165938.4 | XP_005165995.1 | 3 - 4 | 281 | 79 | ||
| XM_005165939.4 | XP_005165996.1 | 3 - 4 | 306 | 79 | ||
| GenBank mRNA | AJ299412.1 | - | 3 - 4 | 219 | 79 | |
| BC051153.1 | - | 3 - 4 | 245 | 79 | ||
| BC155110.1 | - | 3 - 4 | 246 | 79 | ||
More Information
Set Membership: |
|

hdlbpa
Genomic Map