Assay Details
| Gene Symbol: elovl6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201500.1 | NP_958908.1 | 1 - 2 | 142 | 74 | |
| GenBank mRNA | BC046901.1 | - | 1 - 2 | 142 | 74 | |
| BC164614.1 | - | 1 - 2 | 122 | 74 | ||
More Information
Set Membership: |
|

elovl6l
Genomic Map