Assay Details
| Gene Symbol: elovl6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201500.1 | NP_958908.1 | 2 - 3 | 282 | 63 | |
| GenBank mRNA | BC046901.1 | - | 2 - 3 | 282 | 63 | |
| BC164614.1 | - | 2 - 3 | 262 | 63 | ||
More Information
Set Membership: |
|

elovl6l
Genomic Map