Assay Details
| Gene Symbol: mmp13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290479.1 | NP_001277408.1 | 5 - 6 | 835 | 78 | |
| XM_009305556.3 | XP_009303831.1 | 5 - 6 | 835 | 78 | ||
| GenBank mRNA | AF506756.1 | - | 5 - 6 | 808 | 78 | |
| BC049472.1 | - | 5 - 6 | 756 | 78 | ||
| BC065591.1 | - | 5 - 6 | 806 | 78 | ||
| BC159192.1 | - | 5 - 6 | 830 | 78 | ||
More Information
Set Membership: |
|

mmp13a
Genomic Map