Assay Details
| Gene Symbol: mapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201507.1 | NP_958915.1 | 4 - 5 | 870 | 66 | |
| GenBank mRNA | AB030902.1 | - | 4 - 5 | 789 | 66 | |
| AY922319.1 | - | 4 - 5 | 708 | 66 | ||
| BC045505.1 | - | 4 - 5 | 870 | 66 | ||
| BC066401.1 | - | 4 - 5 | 858 | 66 | ||
| BC097073.1 | - | 4 - 5 | 943 | 66 | ||
| BC163960.1 | - | 4 - 5 | 742 | 66 | ||
More Information
Set Membership: |
|

mapk3
Genomic Map