Assay Details
| Gene Symbol: bicc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203420.1 | NP_981965.1 | 17 - 18 | 2349 | 130 | |
| GenBank mRNA | AY513238.1 | - | 17 - 18 | 2349 | 130 | |
More Information
Set Membership: |
|

bicc2
Genomic Map