Assay Details
| Gene Symbol: bicc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203420.1 | NP_981965.1 | 9 - 10 | 1231 | 90 | |
| GenBank mRNA | AY513238.1 | - | 9 - 10 | 1231 | 90 | |
More Information
Set Membership: |
|

bicc2
Genomic Map