Assay Details
| Gene Symbol: uts2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205591.1 | NP_991154.1 | 1 - 2 | 125 | 76 | |
| GenBank mRNA | AY253851.1 | - | 1 - 2 | 125 | 76 | |
| BC162216.1 | - | 1 - 2 | 125 | 76 | ||
| BC162227.1 | - | 1 - 2 | 125 | 76 | ||
More Information
Set Membership: |
|

uts2b
Genomic Map