Assay Details
| Gene Symbol: tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205609.2 | NP_991172.2 | 6 - 7 | 539 | 78 | |
| GenBank mRNA | AY398422.1 | - | 6 - 7 | 519 | 78 | |
| BC115233.1 | - | 6 - 7 | 524 | 78 | ||
| EH494479.1 | - | 6 - 7 | 566 | 78 | ||
More Information
Set Membership: |
|

tbxas1
Genomic Map