Assay Details
| Gene Symbol: dhrs13l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205648.1 | NP_991211.1 | 1 - 2 | 161 | 60 | |
| GenBank mRNA | BC065890.1 | - | 1 - 2 | 161 | 60 | |
More Information
Set Membership: |
|

dhrs13l1
Genomic Map