Assay Details
| Gene Symbol: pdpk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205699.1 | 13 - 14 | 1561 | 73 | ||
| GenBank mRNA | BC065959.1 | - | 13 - 14 | 1561 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdpk1b| Gene Symbol: pdpk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205699.1 | 13 - 14 | 1561 | 73 | ||
| GenBank mRNA | BC065959.1 | - | 13 - 14 | 1561 | 73 | |
Set Membership: |
|