Assay Details
| Gene Symbol: igfbp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126463.1 | NP_001119935.1 | 1 - 2 | 689 | 76 | |
| GenBank mRNA | AY100478.1 | - | 1 - 2 | 685 | 76 | |
| BC066725.1 | - | 1 - 2 | 678 | 76 | ||
More Information
Set Membership: |
|

igfbp5b
Genomic Map