Assay Details
| Gene Symbol: pcdh10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212571.1 | NP_997736.1 | 1 - 2 | 2947 | 97 | |
| XM_009306587.3 | XP_009304862.1 | 1 - 2 | 2991 | 97 | ||
| XM_009306589.3 | XP_009304864.1 | 1 - 2 | 2991 | 97 | ||
| GenBank mRNA | BC066445.1 | - | 1 - 2 | 2947 | 97 | |
More Information
Set Membership: |
|

pcdh10a
Genomic Map