Assay Details
| Gene Symbol: ttpal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212590.2 | NP_997755.2 | 2 - 3 | 480 | 125 | |
| XM_021469732.1 | XP_021325407.1 | 2 - 3 | 515 | 125 | ||
| GenBank mRNA | BC055264.1 | - | 2 - 3 | 469 | 125 | |
| EH434754.1 | - | 2 - 3 | 489 | 125 | ||
More Information
Set Membership: |
|

ttpal
Genomic Map