Assay Details
| Gene Symbol: ywhae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212605.2 | NP_997770.1 | 2 - 3 | 449 | 114 | |
| XM_021481333.1 | XP_021337008.1 | 2 - 3 | 406 | 114 | ||
| GenBank mRNA | BC045325.1 | - | 2 - 3 | 367 | 114 | |
| BC066763.1 | - | 2 - 3 | 350 | 114 | ||
| BC160627.1 | - | 2 - 3 | 354 | 114 | ||
| BC165123.1 | - | 2 - 3 | 300 | 114 | ||
More Information
Set Membership: |
|

ywhae1
Genomic Map