Assay Details
| Gene Symbol: ywhae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212605.2 | NP_997770.1 | 5 - 6 | 896 | 73 | |
| GenBank mRNA | BC045325.1 | - | 5 - 6 | 814 | 73 | |
| BC066763.1 | - | 5 - 6 | 797 | 73 | ||
| BC160627.1 | - | 5 - 6 | 801 | 73 | ||
| BC165123.1 | - | 5 - 6 | 747 | 73 | ||
More Information
Set Membership: |
|

ywhae1
Genomic Map