Assay Details
| Gene Symbol: ahsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212622.2 | NP_997787.2 | 5 - 6 | 781 | 101 | |
| GenBank mRNA | AY217758.2 | - | 5 - 6 | 710 | 101 | |
| BC085588.1 | - | 5 - 6 | 711 | 101 | ||
| BC165731.1 | - | 5 - 6 | 711 | 101 | ||
| EH504898.1 | - | 5 - 6 | 774 | 101 | ||
| EH531076.1 | - | 2 - 3 | 801 | 101 | ||
More Information
Set Membership: |
|

ahsg1
Genomic Map