Assay Details
| Gene Symbol: cd82b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212663.1 | NP_997828.1 | 2 - 3 | 164 | 76 | |
| GenBank mRNA | BC054586.1 | - | 2 - 3 | 164 | 76 | |
| BC164453.1 | - | 1 - 2 | 91 | 76 | ||
More Information
Set Membership: |
|

cd82b
Genomic Map