Assay Details
| Gene Symbol: cd82b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212663.1 | NP_997828.1 | 7 - 8 | 718 | 73 | |
| GenBank mRNA | BC054586.1 | - | 7 - 8 | 718 | 73 | |
| BC164453.1 | - | 6 - 7 | 645 | 73 | ||
More Information
Set Membership: |
|

cd82b
Genomic Map