Assay Details
| Gene Symbol: brd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212702.1 | NP_997867.1 | 6 - 7 | 1223 | 95 | |
| XM_005165593.4 | XP_005165650.2 | 6 - 7 | 1289 | 95 | ||
| XM_005165594.4 | XP_005165651.2 | 6 - 7 | 1272 | 95 | ||
| XM_005165595.4 | XP_005165652.2 | 6 - 7 | 1275 | 95 | ||
| XM_009301936.3 | XP_009300211.1 | 6 - 7 | 1275 | 95 | ||
| XM_009301938.3 | XP_009300213.1 | 6 - 7 | 1275 | 95 | ||
| XM_009301940.3 | XP_009300215.1 | 6 - 7 | 1275 | 95 | ||
| GenBank mRNA | BC055533.1 | - | 6 - 7 | 1289 | 95 | |
| BC065949.1 | - | 6 - 7 | 1223 | 95 | ||
| BC151886.1 | - | 6 - 7 | 1257 | 95 | ||
| BC152644.1 | - | 6 - 7 | 1224 | 95 | ||
More Information
Set Membership: |
|

brd3b
Genomic Map