Assay Details
| Gene Symbol: brd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212702.1 | NP_997867.1 | 8 - 9 | 1539 | 114 | |
| XM_005165595.4 | XP_005165652.2 | 8 - 9 | 1591 | 114 | ||
| XM_009301938.3 | XP_009300213.1 | 8 - 9 | 1591 | 114 | ||
| GenBank mRNA | BC065949.1 | - | 8 - 9 | 1539 | 114 | |
| BC152644.1 | - | 8 - 9 | 1540 | 114 | ||
More Information
Set Membership: |
|

brd3b
Genomic Map