Assay Details
| Gene Symbol: kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212742.1 | NP_997907.1 | 4 - 5 | 633 | 87 | |
| GenBank mRNA | BC055629.1 | - | 4 - 5 | 633 | 87 | |
| BC155277.1 | - | 4 - 5 | 683 | 87 | ||
| BC171589.1 | - | 4 - 5 | 623 | 87 | ||
More Information
Set Membership: |
|

kat8
Genomic Map