Assay Details
| Gene Symbol: rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243037.2 | NP_001229966.1 | 4 - 5 | 261 | 73 | |
| NM_001319650.1 | NP_001306579.1 | 4 - 5 | 430 | 73 | ||
| GenBank mRNA | BC055097.1 | - | 3 - 4 | 222 | 73 | |
| CF996097.1 | - | 3 - 4 | 273 | 73 | ||
| EB874078.1 | - | 4 - 5 | 252 | 73 | ||
| EH488885.1 | - | 4 - 5 | 304 | 73 | ||
| EH543457.1 | - | 4 - 5 | 291 | 73 | ||
More Information
Set Membership: |
|

rpl17
Genomic Map