Assay Details
| Gene Symbol: ypel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212790.1 | NP_997955.1 | 4 - 5 | 314 | 71 | |
| GenBank mRNA | BC050961.1 | - | 4 - 5 | 572 | 71 | |
| BC067578.1 | - | 4 - 5 | 314 | 71 | ||
| BC154300.1 | - | 4 - 5 | 389 | 71 | ||
More Information
Set Membership: |
|

ypel3
Genomic Map