Assay Details
| Gene Symbol: tlr4bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212813.2 | NP_997978.2 | 1 - 2 | 277 | 102 | |
| GenBank mRNA | AY388400.1 | - | 1 - 2 | 81 | 102 | |
| BC068358.1 | - | 1 - 2 | 270 | 102 | ||
More Information
Set Membership: |
|

tlr4bb
Genomic Map