Assay Details
| Gene Symbol: b3gnt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212840.1 | NP_998005.1 | 1 | 1452 | 125 | |
| XM_005160144.4 | XP_005160201.1 | 3 | 2341 | 125 | ||
| XM_005160145.4 | XP_005160202.1 | 3 | 2361 | 125 | ||
| XM_005160146.4 | XP_005160203.1 | 2 | 2247 | 125 | ||
| GenBank mRNA | AF321828.1 | - | 1 | 1452 | 125 | |
More Information
Set Membership: |
|

b3gnt2a
Genomic Map