Assay Details
| Gene Symbol: yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212891.1 | NP_998056.1 | 3 - 4 | 516 | 145 | |
| GenBank mRNA | BC066619.1 | - | 3 - 4 | 516 | 145 | |
More Information
Set Membership: |
|

yipf4
Genomic Map