Assay Details
| Gene Symbol: chmp2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212904.2 | NP_998069.1 | 2 - 3 | 256 | 80 | |
| GenBank mRNA | BC067142.1 | - | 2 - 3 | 210 | 80 | |
More Information
Set Membership: |
|

chmp2bb
Genomic Map