Assay Details
| Gene Symbol: pdia7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212905.1 | NP_998070.1 | 7 - 8 | 899 | 64 | |
| GenBank mRNA | BC067155.1 | - | 7 - 8 | 899 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdia7| Gene Symbol: pdia7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212905.1 | NP_998070.1 | 7 - 8 | 899 | 64 | |
| GenBank mRNA | BC067155.1 | - | 7 - 8 | 899 | 64 | |
Set Membership: |
|