Assay Details
| Gene Symbol: ucmab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212934.2 | NP_998099.1 | 3 - 4 | 300 | 63 | |
| GenBank mRNA | BC067652.1 | - | 3 - 4 | 265 | 63 | |
| JQ003911.1 | - | 3 - 4 | 287 | 63 | ||
More Information
Set Membership: |
|

ucmab
Genomic Map