Assay Details
| Gene Symbol: lnx2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212940.2 | NP_998105.2 | 2 - 3 | 738 | 72 | |
| XM_005173252.3 | XP_005173309.1 | 3 - 4 | 894 | 72 | ||
| XM_005173253.4 | XP_005173310.1 | 2 - 3 | 752 | 72 | ||
| GenBank mRNA | BC139538.1 | - | 2 - 3 | 738 | 72 | |
| FJ156084.1 | - | 2 - 3 | 725 | 72 | ||
More Information
Set Membership: |
|

lnx2b
Genomic Map