Assay Details
| Gene Symbol: lnx2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212940.2 | NP_998105.2 | 4 - 5 | 1122 | 74 | |
| XM_005173252.3 | XP_005173309.1 | 5 - 6 | 1278 | 74 | ||
| XM_005173253.4 | XP_005173310.1 | 4 - 5 | 1136 | 74 | ||
| GenBank mRNA | BC139538.1 | - | 4 - 5 | 1122 | 74 | |
| FJ156084.1 | - | 4 - 5 | 1109 | 74 | ||
More Information
Set Membership: |
|

lnx2b
Genomic Map