Assay Details
| Gene Symbol: lnx2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212940.2 | NP_998105.2 | 6 - 7 | 1662 | 95 | |
| XM_005173252.3 | XP_005173309.1 | 7 - 8 | 1818 | 95 | ||
| XM_005173253.4 | XP_005173310.1 | 6 - 7 | 1676 | 95 | ||
| GenBank mRNA | BC139538.1 | - | 6 - 7 | 1662 | 95 | |
| FJ156084.1 | - | 6 - 7 | 1649 | 95 | ||
More Information
Set Membership: |
|

lnx2b
Genomic Map